Description
The Thermo Scientific Dionex UltiMate 3000 HPLC system is applied for the separation of glycerides. The analysis demonstrates an alternative use of the Thermo Scientific Acclaim HILIC-10 HPLC column, operating in normal-phase (NP) mode. This gradient flow method allows for a quick and simple separation of triglycerides, diglycerides and mononogycerides with charged aerosol detection (CAD).| Market: | Food and Beverage |
| Keywords: | Diglycerides, Triglycerides, Charged aerosol detection, Glycerides, Mononogycerides, Acclaim HILIC-10 |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.