Find methods for your needs
Municipal Wastewater Analysis According to EPA Method 300.1
Description
Anion determinations in municipal waste water samples using EPA Method 300.1 on an integrated IC System with a Thermo Scientific Dionex IonPac AS22 anion-exchange column to separate and quantify inorganic anions with manually-prepared carbonate/bicarbonate eluents on a Thermo Scientific Dionex ICS-1100 or a Thermo Scientific Dionex ICS-1600 IC system. This method provides an economical way to meet EPA Method 300.1 (A) regulatory testing requirements.| Market: | Environmental |
| Matrix: | Municipal wastewater |
For Research Use Only. Not for use in diagnostic procedures.