Find methods for your needs
Ultrafast analysis of Aspirin by UHPLC using a Thermo Scientific Acclaim 120 C18 2.2 µm HPLC column
Description
The Thermo Scientific UltiMate 3000 RSLC system is applied for the rapid determination of aspirin. The separation is performed on a Thermo Scientific Acclaim 120 C18 2.2 µm column in under 1.5 mins, with UV detection at 280 nm. This application is a speed-up of the USP method for the assay of Aspirin. Simply recomputing the operating parameters for a 2.1x50 mm column leads to a 10-fold increase in throughput and 96% savings in mobile phase per assay, whilst maintaining baseline separation.Market: | Pharma |
Keywords: | Acclaim 120 C18, Aspirin, HPLC, Impurities, Salicylic acid, USP, UV detection |
Matrix: | Tablet |
For Research Use Only. Not for use in diagnostic procedures.