Find methods for your needs
Rapid analysis of vitamin D3 in supplements using the Thermo Scientific Acclaim HILIC-10 HPLC column
Description
The Thermo Scientific Dionex UltiMate 3000 RSLC system is applied for the rapid analysis of vitamin D3 in oil based vitamin supplements. The separation is performed isocratically on a Thermo Scientific Acclaim HILIC-10 column, with UV detection at 265 nm.| Market: | Pharma |
| Keywords: | Acclaim HILIC-10, HPLC, UV detection, Vitamin D3, Vitamin supplement |
| Matrix: | Vitamin supplement |
For Research Use Only. Not for use in diagnostic procedures.