Find methods for your needs
Enhanced separation of keto acids using the Thermo Scientific Acclaim Mixed-Mode WAX-1 HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of small organic acids. The separation is performed isocratically on a Thermo Scientific Acclaim Mixed-Mode WAX-1, with UV detection at 210 nm.Market: | Chemical |
Keywords: | 2-ketobutyrate, 2-ketoglutarate, 2-ketovalerate, Acclaim Mixed-Mode WAX-1, HPLC, Organic acids, Pyruvate, UV detection |
Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.