Find methods for your needs
Improved analysis of polyethylene glycols using a Thermo Scientific Acclaim SEC-300 HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of polyethylene glycols (PEGs) by size exclusion chromatography. The separation is performed on a Thermo Scientific Acclaim SEC-300 HPLC column, with RI detection. The separation achieved is compared to the separation obtained on a Thermo Scientific Acclaim SEC-1000 HPLC column. Both give excellent performance for measuring molecular weight and size distribution for this class of polymers.| Market: | Chemical |
| Keywords: | Acclaim SEC-300, HPLC, Polyethylene glycol (PEG), Refractive index detection, Size exclusion |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.