Find methods for your needs
High resolution separation of polyethylene glycols (PEGs) using the Thermo Scientific Acclaim Surfactant Plus HPLC column with charged aeorosol detection
Description
The Thermo Scientific Acclaim Surfactant Plus HPLC column provides excellent resolution for the low molecular weight oligomers based on the degree of ethoxylation. The optimized pore size allows it to resolve polymers up to n = 30.| Market: | Chemical |
| Keywords: | Acclaim Surfactant Plus, Charged aerosol detection, HPLC, Polyethylene glycol (PEG) |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.