Find methods for your needs
Excellent separation of human IgG glycans on a Thermo Scientific Accucore 150-Amide-HILIC HPLC column
Description
Human IgG glycans were separated on a Thermo Scientific Accucore 150-Amide-HILIC HPLC column using 2-AB derivatization and fluorescence detection.Market: | BioPharma |
Keywords: | Accucore 150-Amide-HILIC, Fluorescence detection, glycans, HPLC, Human IgG |
Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.