Find methods for your needs
Fast analysis of beta blockers using a Thermo Scientific Accucore RP-MS HPLC column
Description
The Thermo Scientific HPLC system is applied for the determination of six beta blockers. The separation is performed on a Thermo Scientific Accucore RP-MS HPLC column, with UV detection at 220 nm.| Market: | Pharma |
| Keywords: | Accucore RP-MS, Alprenolol, Atenolol, Beta blockers, HPLC, Metoprolol, Nadolol, Pindolol, Propranolol, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.