Find methods for your needs
Improved analysis of vitamin K isomers using a Thermo Scientific Accucore C30 HPLC column
Description
The Thermo Scientific HPLC system is applied for the determination of vitamin K isomers. The separation is performed on a Thermo Scientific Accucore C30 HPLC column, with UV detection at 250 nm.| Market: | Food and Beverage |
| Keywords: | Accucore C30, HPLC, Isomers, UV detection, Vitamin K |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.