Find methods for your needs
Fast analysis of norethindrone and mestranol with progesterone as internal standard using a Thermo Scientific Syncronis C8 HPLC column
Description
The Thermo Scientific UltiMate 3000 system is applied for the determination of norethindrone and mestranol with progesterone as internal standard. The separation is performed on a Thermo Scientific Syncronis C8 HPLC column, with UV detection at 200 nm.| Market: | Pharma |
| Keywords: | HPLC, Mestranol, Norethindrone, Progesterone, Syncronis C8, UltiMate 3000, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.