Find methods for your needs
Rapid LC-MS/MS analysis of Montelukast in human plasma using Thermo Scientific SOLA SAX SPE and a Thermo Scientific Accucore C8 HPLC column
Description
The Thermo Scientific Dionex UltiMate 3000 HPLC system is applied for the analysis of montelukast in human plasma. The samples were prepared using a SOLA SAX solid phase extraction (SPE) 96 well plate, and the separation performed in under 1.5 minutes on a Thermo Scientific Accucore C8 HPLC column, with MS/MS detection. The dynamic range was linear between 1 and 1000ng/mL with a R² of 0.9901. For Research Use Only. Not for use in diagnostic procedures.| Market: | Pharma |
| Keywords: | Accucore C8, LC-MS/MS, Montelukast, MS detection, SOLA SAX, SPE |
| Matrix: | Human plasma |
For Research Use Only. Not for use in diagnostic procedures.