Find methods for your needs
Rapid determination of Rosuvastatin in human plasma by SPE-LC-MS/MS using Thermo Scientific SOLA HRP SPE and a Thermo Scientific Accucore RP-MS HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of rosuvastatin in human plasma. The samples were prepared using a SOLA HRP solid phase extraction (SPE) 96 well plate, and the separation performed in under 3.6 minutes on a Thermo Scientific Accucore RP-MS HPLC column, with MS/MS detection. For Research Use Only. Not for use in diagnostic procedures.| Market: | Pharma |
| Keywords: | LC-MS/MS, MS detection, Plasma, Rosuvastatin, SOLA, SPE |
| Matrix: | Human plasma |
For Research Use Only. Not for use in diagnostic procedures.