Find methods for your needs
Improved analysis of tetracyclines using a Thermo Scientific Hypersil GOLD HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of tetracyclines. The separation is performed on a Thermo Scientific Hypersil GOLD HPLC column, with UV detection at 350 nm.| Market: | Pharma |
| Keywords: | Doxycycline, Epi-tetracycline, HPLC, Hypersil GOLD, Methacycline, Oxytetracycline, Tetracycline, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.