Find methods for your needs
Rapid determination of prednisone using a Thermo Scientific Accucore C18 HPLC column and MS detection
Description
A simple, rapid and sensitive procedure for the determination of prednisone by liquid chromatography-tandem mass spectrometry was developed and evaluated. The drug was separated on an Accucore C18 HPLC column under reversed-phase, gradient conditions. Detection was performed on a triple quadrupole mass spectrometer using positive polarity, heated electrospray ionisation (HESI) conditions operating in selected reaction monitoring (SRM) mode.| Market: | Pharma |
| Keywords: | Accucore C18, HPLC, MS detection, Prednisone |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.