Find methods for your needs
Simple and rapid method for the determination of asenapine maleate impurities using a solid core C8 HPLC column
Description
The Thermo Scientific Dionex UltiMate 3000 HPLC system is applied for the determination of asenapine maleate impurities. The separation is performed on a Thermo Scientific Accucore C8 HPLC column which provides excellent resolution (Rs >12) between asenapine and its cis-isomer impurity.| Market: | Pharma |
| Keywords: | Accucore C8, Asenapine maleate, HPLC, Impurities, UV detection |
| Matrix: | N/A |
| Author: | Prakash Chander |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.