Find methods for your needs
Improved analysis of steroids using a Thermo Scientific Hypersil GOLD CN HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of steroids. The separation is performed on a Thermo Scientific Hypersil GOLD CN HPLC column, with UV detection at 254 nm.| Market: | Pharma |
| Keywords: | Corticosterone, Cortisone, Deoxycorticosterone, HPLC, Hydrocortisone, Hydroxyprogesterone, Hypersil GOLD CN, Progesterone, Steroids, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.