Find methods for your needs
Fast analysis of banned aromatic amines using a Thermo Scientific Hypersil GOLD PFP HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of banned aromatic amines. The separation is performed on a Thermo Scientific Hypersil GOLD PFP 1.9 µm HPLC column, with UV detection at 254 nm.| Market: | Environmental |
| Keywords: | Aromatic amines, Diaminotoluene, HPLC, Hypersil Gold PFP, Oxydianiline, Toluidine, Trimethylaniline, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.