Find methods for your needs
Fast analysis of ketones using a Thermo Scientific Accucore C18 HPLC column
Description
The Thermo Scientific HPLC system is applied for the determination of ketones. The separation is performed on a Thermo Scientific Accucore C18 HPLC column in less than 2 minutes, with UV detection at 258 nm.| Market: | Chemical |
| Keywords: | Accucore C18, Acetophenone, Butyrophenone, Decanophenone, Heptanone, Hexanophenone, HPLC, Ketones, Octanophenone, Pentanone, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.