Find methods for your needs
Fast analysis of banned aromatic amines using a Thermo Scientific Hypersil GOLD HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of banned aromatic amines. The separation is performed on a Thermo Scientific Hypersil GOLD 1.9 µm HPLC column, with UV detection at 254 nm.| Market: | Environmental |
| Keywords: | 2,4,5-Trimethylaniline, 2,4-Diaminotoluene, 2-Methoxy-5-Methylaniline, 4,4-Methylene-bis(2-chloroaniline), 4,4-Oxydianiline, HPLC, Hypersil GOLD, O-Toluidine, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.