Find methods for your needs
Rapid analysis of rosuvastatin using a Thermo Scientific Syncronis C18 HPLC column Be the first to rate this application
Description
The Thermo Scientific HPLC system is applied for the determination of rosuvastatin. The separation is performed on a Thermo Scientific Syncronis C18 HPLC column, with UV detection at 242 nm.
Market: Pharma
Keywords: HPLC, Rosuvastatin, Syncronis C18, UV detection
Matrix: N/A
Uploaded on 8/31/2012.
For Research Use Only. Not for use in diagnostic procedures.