Find methods for your needs
Improved analysis of catecholamines using a Thermo Scientific Hypersil GOLD aQ HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of catecholamines. The separation is performed on a Thermo Scientific Hypersil GOLD aQ HPLC column, with UV detection at 270 nm.| Market: | Pharma |
| Keywords: | 5-HIAA, 5-Hydroxydopamine, Catecholamines, DOPAC, Dopamine, HPLC, Hypersil GOLD aQ, Serotonin, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.