Find methods for your needs
Fast analysis of alkaloids using a Thermo Scientific Hypersil GOLD aQ HPLC column Be the first to rate this application
Description
The Thermo Scientific HPLC system is applied for the analysis of alkaloids. The separation is performed on a Thermo Scientific Hypersil GOLD aQ HPLC column, with UV detection at 260 nm.
Market: Chemical
Keywords: Alkaloids, Anabasine, Cotinine, HPLC, Hypersil GOLD aQ, Nicotine, UV detection
Matrix: N/A
Uploaded on 8/31/2012.
For Research Use Only. Not for use in diagnostic procedures.