Find methods for your needs
Improved analysis of Folic Acid using a Thermo Scientific Hypersil GOLD HPLC column Be the first to rate this application
improved analysis folic acid using a thermo scientific hypersil gold hplc column
Description
The Thermo Scientific HPLC system is applied for the analysis of folic acid using the USP method. The separation is performed on a Thermo Scientific Hypersil GOLD HPLC column, with UV detection at 268 nm.
Market: Food and Beverage
Keywords: Folic acid, HPLC, Hypersil GOLD, USP, UV detection
Matrix: N/A
Uploaded on 8/30/2012.
For Research Use Only. Not for use in diagnostic procedures.