Find methods for your needs
Fast analysis of zidovudine by UHPLC using a Thermo Scientific Syncronis C18 1.7 µm column
Description
The Thermo Scientific HPLC system is applied for the determination of zidovudine. The separation is performed on a Thermo Scientific Syncronis C18 UHPLC column, with UV detection at 265 nm.Market: | Pharma |
Keywords: | HPLC, Syncronis C18, UV detection, Zidovudine |
Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.