Find methods for your needs
Rapid LC-MS/MS method for the determination of Docetaxel in human serum using Thermo Scientific SOLA HRP SPE and a Thermo Scientific Accucore RP-MS HPLC column
Description
The Thermo Scientific Dionex UltiMate 3000 RSLC system is applied for the analysis of Docetaxel in human serum. The sample was prepared using SOLA HRP solid phase extraction (SPE) 96 well plates, and the separation performed on a Thermo Scientific Accucore RP-MS HPLC column in less than 3 minutes, with MS/MS detection. For Research Use Only. Not for use in diagnostic procedures.| Market: | Pharma |
| Keywords: | Accucore RP-MS, Chemotherapy drug, Docetaxel, Human serum, LC-MS/MS, MS detection, SOLA HRP, SPE |
| Matrix: | Human serum |
For Research Use Only. Not for use in diagnostic procedures.