Find methods for your needs
Rapid determination of esomeprazole using a Thermo Scientific Syncronis C18 1.7 μm UHPLC column Be the first to rate this application
rapid determination esomeprazole using a thermo scientific syncronis c18 17 μm uhplc column
Description
The Thermo Scientific HPLC system is applied for the determination of esomeprazole. The separation is performed on a Thermo Scientific Syncronis C18 UHPLC column, with UV detection at 302 nm.
Market: Pharma
Keywords: Esomeprazole, HPLC, Syncronis C18, UV detection
Matrix: N/A
Uploaded on 8/30/2012.
For Research Use Only. Not for use in diagnostic procedures.