Find methods for your needs
Improved analysis of Linoleic acid using a Thermo Scientific Hypersil GOLD C8 HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of linoleic acid. The separation is performed on a Thermo Scientific Hypersil GOLD C8 HPLC column, with UV detection at 200 nm.| Market: | Chemical |
| Keywords: | FAMEs, Fatty acid methyl esters, HPLC, Hypersil GOLD C8, Linoleic acid, Linolenic acid, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.