Find methods for your needs
Improved analysis of triazines using a Thermo Scientific Hypersil GOLD C8 HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of triazines. The separation is performed on a Thermo Scientific Hypersil GOLD C8 HPLC column, with UV detection at 220 nm.| Market: | Environmental |
| Keywords: | Ametryn, Atrazine, Herbicides, HPLC, Hypersil GOLD C8, Prometon, Prometryn, Propazine, Simazine, Simetryn, Triazines, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.