Find methods for your needs
Fast analysis of procainamides using a Thermo Scientific Hypersil GOLD HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of procainamides. The separation is performed on a Thermo Scientific Hypersil GOLD HPLC column, with UV detection at 254 nm.| Market: | Pharma |
| Keywords: | Acetyl-Procainamide, Caffeine, HPLC, Hypersil GOLD, Procainamide, Procainamides, Propionyl procainamide, Uracil, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.