Find methods for your needs
Fast analysis of urea herbicides using a Thermo Scientific Hypersil GOLD C8 HPLC column Be the first to rate this application
Description
The Thermo Scientific HPLC system is applied for the analysis of urea herbicides. The separation is performed on a Thermo Scientific Hypersil GOLD C8 HPLC column, with UV detection at 240 nm.
Market: Environmental
Keywords: Chlorotoluron, Diuron, HPLC, Hypersil GOLD C8, Linuron, Metoxuron, Monuron, Tebuthiuron, Urea herbicides, UV detection
Matrix: N/A
Uploaded on 8/29/2012.
For Research Use Only. Not for use in diagnostic procedures.