Find methods for your needs
Fast analysis of chloramphenicol and thiamphenicol using reversed-phase HPLC-UV
Description
The Thermo Scientific HPLC system is applied for the analysis of antibacterials. The separation is performed on a Thermo Scientific Hypersil GOLD HPLC column, with UV detection at 240 nm.| Market: | Pharma |
| Keywords: | Antibacterials, Chloramphenicol, HPLC, Hypersil GOLD, Thiamphenicol, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.