Find methods for your needs
Improved analysis of Lidocaine and preservatives using a Thermo Scientific Hypersil GOLD C8 HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of parabens. The separation is performed on a Thermo Scientific Hypersil GOLD C8 HPLC column, with UV detection at 230 nm.| Market: | Pharma |
| Keywords: | HPLC, Hypersil GOLD C8, Lidocaine, Methyl paraben, Paraben, Preservatives, Propyl paraben, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.