Find methods for your needs
Improved analysis of triazine and uron herbicides using a Thermo Scientific Hypersil GOLD C8 HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of triazine and uron herbicides. The separation is performed on a Thermo Scientific Hypersil GOLD C8 HPLC column, with UV detection at 240 nm.| Market: | Environmental |
| Keywords: | Atrazine, Chlorotoluron, Diuron, HPLC, Hypersil GOLD C8, Linuron, Monuron, Propazine, Simazine, Triazine herbicides, Uron herbicides, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.