Find methods for your needs
Improved analysis of antibiotics using a Thermo Scientific Hypersil GOLD HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of antibiotics. The separation is performed on a Thermo Scientific Hypersil GOLD HPLC column, with UV detection at 254 nm.| Market: | Pharma |
| Keywords: | Cefaclor, Cefadroxil, Cefazolin, Cephalexin, Cephalosporin Antibiotic, Cephradine, HPLC, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.