Find methods for your needs
Improved analysis of coumaric acids using a Thermo Scientific Hypersil GOLD HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of coumaric acids. The separation is performed on a Thermo Scientific Hypersil GOLD HPLC column, with UV detection at 270 nm.| Market: | Pharma |
| Keywords: | Coumaric acid, HPLC, Hypersil GOLD, Uracil, UV detection |
| Matrix: | Standard |
For Research Use Only. Not for use in diagnostic procedures.