Find methods for your needs
Fast analysis of a lorazepam tablet using a Thermo Scientific Hypersil GOLD HPLC column Be the first to rate this application
Description
The Thermo Scientific HPLC system is applied for the analysis of a lorazepam tablet. The separation is performed on a Thermo Scientific Hypersil GOLD HPLC column in less than 5 minutes, with UV detection at 235 nm.
Market: Pharma
Keywords: HPLC, Hypersil GOLD, Lorazepam, Tablet, UV detection
Matrix: Tablet
Uploaded on 8/28/2012.
For Research Use Only. Not for use in diagnostic procedures.