Find methods for your needs
Fast determination of Zolmitriptan in human serum by LC-MS/MS using Thermo Scientific SOLA CX SPE and a Thermo Scientific Hypersil GOLD HPLC column
Description
The Thermo Scientific Dionex UltiMate 3000 HPLC system is applied for the analysis of Zolmitriptan in human serum. The sample was prepared using SOLA CX solid phase extraction (SPE) cartridges, and the separation performed on a Thermo Scientific Hypersil GOLD HPLC column in less than 2.5 minutes, with MS/MS detection. For Research Use Only. Not for use in diagnostic procedures.| Market: | BioPharma |
| Keywords: | Diphenhydramine, Human serum, Hypersil GOLD, LC-MS/MS, MS detection, Serotonin Receptor Agonist, SOLA CX, SPE, Zolmitriptan |
| Matrix: | Human serum |
For Research Use Only. Not for use in diagnostic procedures.