Find methods for your needs
AN72333: Determination of Adsorbable Organic Halogen in Wastewater using a Combustion Ion Chromatography System
Description
This application note demonstrates that AOX can be precisely and accurately determined in wastewater using combustion ion chromatography. Analysis was automated using the Mitsubishi AQF–2100H system in combination with the Dionex Integrion HPIC system with a Dionex IonPac AS18-4μm column.| Market: | Water Analysis |
| Keywords: | Dionex IonPac AS18-4μm Column, HPIC, CIC, AQF-2100H, RFIC System, Wastewater analysis, Dionex Integrion HPIC System, TN72211, AN 72333, Organic halogen |
| Matrix: | Waste water |
| Author: | Jingli Hu; Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.