Find methods for your needs
Tandem Operation of gradient methods for water soluble vitamins
Description
Many fast UHPLC and traditional HPLC methods suffer under delays caused by the need to wash and equilibrate the column after the separation. Typically we see 20%–40% of the runtime used up by these essential steps within a gradient method. Here a solution utilizing a dual-gradient pump setup with column switching capabilities to overcome these delays without the need of redeveloping existing methods is offered. The Thermo Scientific™ Dionex™ UltiMate™ 3000 LC Dual System with the Viper Tandem Operation kit allows to sppeding up gradients in LC and UHPLC assays.| Market: | Pharma |
| Keywords: | HPLC, Water soluble vitamins, UHPLC, ×2 Dual, Dual-Gradient, Gradient Method, Tandem Operation, UltiMate 3000 LC System |
| Status: | Proof of Concept |
For Research Use Only. Not for use in diagnostic procedures.