Find methods for your needs
AN327: Extraction of Nitroglycerin from Transdermal Patches by Accelerated Solvent Extraction
Description
Accelerated solvent extraction is an extraction method that significantly streamlines sample preparation. Solvent is pumped into an extraction cell containing the sample, which is then brought to an elevated temperature and pressure. The entire extraction process is fully automated and performed in minutes for fast and easy extraction with low solvent consumption. The work reported here was undertaken to investigate the use of accelerated solvent extraction for the extraction of nitroglycerin from prototype transdermal matrix patches.| Market: | Pharma |
| Keywords: | ASE, ASE 200, ASE 350, Transdermal Patches, Nitroglycerin |
| Matrix: | Drug product |
For Research Use Only. Not for use in diagnostic procedures.