Find methods for your needs
AN350: Extraction of Zearalenone from Wheat and Corn by Accelerated Solvent Extraction (ASE)
Description
Zearalenone (ZON) is a mycotoxin produced by the Fusarium fungus. ZON can be found in a wide variety of plants and soils, and can have negative health effects on animal husbandry and humans. Accelerated Solvent Extraction (ASE) is a proven extraction technology that not only helps to eliminate these bottlenecks by decreasing the extraction time, but requires far less technician attendance because it is an automated system.| Market: | Food and Beverage |
| Keywords: | ASE, ASE 200, ASE 350, Zearalenone (ZON), Mycotoxin, Plants, Grains |
For Research Use Only. Not for use in diagnostic procedures.