Find methods for your needs
AN1004: Automated Solid-Phase Extraction of Organochlorine Pesticides from Drinking Water
Description
Twenty chlorinated pesticides were extracted from drinking water using a Thermo Scientific Dionex AutoTrace 280 Solid Phase Extraction Instrument (AT 280). The efficiency of the Dionex AT280 method was compared to the traditional liquid-liquid phase extraction (LLE). The manual extraction required significantly more time compared to the Dionex AutoTrace 280 instrument. Analyte recovery was improved using the AT280 Automated SPE instrument for the majority of compounds without sacrificing the precision of analysis.| Market: | Food and Beverage |
| Keywords: | Organochlorine pesticides, Heptachlor, Heptachlor epoxide, trans-Chlordane, Methoxychlor, Aldrin, Cis-Chlordane, Endosulfan I, Dieldrin, Endrin, Endrin Aldehyde, Endosulfan sulfate, Endrin ketone, Drinking water, AT 280, SolEx C-18 Extraction Columns, Alpha-BHC, Gamma-BHC, Beta-BHC, Delta-BHN, 4, 4’-DDE, 4-4’-DDD, Endosulfate II, 4, 4’-DDT |
| Matrix: | Drinking water |
| Author: | Selvan Lingam, Bruce Richter, Richard Calson, Brett Murphy |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.