Find methods for your needs
AN1025: Simultaneous Extraction of PAHs and PCBs from Environmental Samples Using Accelerated Solvent Extraction
Description
Accelerated solvent extraction is an established technique to efficiently extract contaminants in solid and semisolid sample matrices with less solvent and time. Polyaromatic Hydrocarbons (PAHs) and Polychlorinated Biphenyls (PCBs) are considered toxic and carcinogenic, and therefore classified as persistent organic pollutants (POPs). Typically, PAHs and PCBs are extracted separately, using different solvent combinations. Our study demonstrates extraction of PAHs and PCBs using a single method from spiked mussel and from a soil spiked with a standard reference materials (SRMs).| Market: | Environmental |
| Keywords: | Acenaphthene, Acenaphthylene, Anthracene, Chrysene, Fluoranthene, Fluorene, Naphthalene, PAHs, Phenanthrene, Polychlorinated biphenyls (PCBs), Pyrene, Accelerated solvent extraction, ASE, Polyaromatic hydrocarbons, Polychlorinated biphenyls, ASE 200, ASE 350, PCBs, U.S. EPA Method 3545A, Polyaromatic Hydrocarbons (PAHs), Aroclor 1254, tetrachloro xylene, 2,4,5,6-tetrachloro-m-xylene, Indeno(1,2,3-cd) pyrene, Indenopyrene, Dibenzo anthracene, Benzoperylene, Benzopyrene, Benzofluoranthene, Benzoanthracene, p-Terphenyl-d4, 2-Fluorobiphenyl, Nitrobenzene-d5 |
| Matrix: | Green lipped mussels, Soil |
| Author: | Brett Murphy, Selvan Lingam, Bruce Richter, and Richard Carlson |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.