Find methods for your needs
AN317: Extraction of BNAs (Bases, Neutrals, and Acids) Using Accelerated Solvent Extraction (ASE)
Description
The procedures described in this application note meet the requirements for sample extraction as determined by U.S. EPA Method 3545. This method is applicable to the extraction of water-insoluble or slightly water-soluble volatile and semivolatile compounds in preparation for gas chromatographic or GC/MS measurement. This method is applicable to soils, clays, wastes, and sediments containing 250–12,500 μg/kg of BNA compounds.| Market: | Environmental |
| Keywords: | soil, ASE 200, ASE 350, BNA, EPA SW-846 method 3545, ASE Prep DE, Clay, Sand, Loam |
| Matrix: | Soil |
For Research Use Only. Not for use in diagnostic procedures.