Find methods for your needs
Related substances analysis of cefepime dihydrochloride hydrate using a C18 HPLC column as per JP XVI and KP X monograph method
Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied for the related substances analysis of cefepime dihydrochloride hydrate. The analysis was performed on a Thermo Scientific Syncronis aQ HPLC column using the method described in the JP XVI and KP X. The results obtained for theoretical plates and %RSD for cefepime met the criteria stated in the JP/KP monograph. The results obtained for theoretical plates of cefepime was 54,132 (JP/KP criteria of not less than 6,000) and %RSD for cefepime was 0.1% (JP/KP criteria of not more than 2).| Market: | Pharma |
| Keywords: | Cephalosporin Antibiotic, HPLC, UV detection, JP XVI monograph, KP X monograph, Cefepime dihydrochloride |
| Matrix: | N/A |
| Author: | Krishnarao Dara |
| Affiliation: | Thermo Fisher Scientific, APAC CoE Lab, Ahmedabad, India |
For Research Use Only. Not for use in diagnostic procedures.