Find methods for your needs
LC-MS/MS analysis of polar pesticides in honey with QuPPe extraction
Description
We present a sensitive, robust, fast method to determine and quantify residues of polar pesticides in food samples in complex matrices at required regulatory levels. The Thermo Scientific UltiMate 3400 RSLC system is used in combination with a Thermo Scientific TSQ Quantiva triple quadrupole mass spectrometer to analyze QuPPe method extracts. The robustness of the system allows uninterrupted analysis of up to 500 honey samples and easy cleaning of the ion introduction system.| Market: | Food and Beverage |
| Keywords: | AMPA, Glyphosate, Hypercarb, TSQ Quantiva, polar pesticides, QuPPe |
| Matrix: | Honey |
| Author: | Claudia Martins and Michal Godula |
| Affiliation: | Thermo FIsher Scientific |