Find methods for your needs
Rapid analysis of six aromatic amines using a Thermo Scientific Accucore PFP HPLC column Be the first to rate this application
Description
The Thermo Scientific HPLC system is applied for the analysis of six aromatic amines. The separation is performed on a Thermo Scientific Accucore PFP HPLC column in less than 2 minutes, with UV detection at 254 nm.
Market: Chemical
Keywords: 2,4,5-Trimethylaniline, 2,4-Diaminotoluene, 2-Methyoxy-5-methylaniline, 4,4-Methylene-bis, 4,4-Oxydianiline, Accucore PFP, Aromatic amines, HPLC, O-Toluidine, UV detection
Matrix: N/A
Uploaded on 8/23/2012.
For Research Use Only. Not for use in diagnostic procedures.