Find methods for your needs
Fast analysis of steroids using a Thermo Scientific Accucore RP-MS HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of steroids. The separation is performed on a Thermo Scientific Accucore RP-MS HPLC column in less than 6 minutes, with UV detection at 254 nm.| Market: | Pharma |
| Keywords: | 11-alpha Hydroxyprogesterone, 17-alpha Hydroxyprogesterone, Accucore RP-MS, Corticosterone, Cortisone, Hormones, HPLC, Hydrocortisone, Progesterone, Steroids, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.