Find methods for your needs
Separation of related impurities of Methocarbamol Injection.
Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied for the separation of related impurities of Methocarbamol Injection. The separation was performed on a Thermo Scientific Acclaim Phenyl-1 HPLC column with UV detection at 276 nm. With a total run time of 65 minutes the method allows separation of all related impurities of Methocarbamol.| Market: | Pharma |
| Keywords: | Acclaim Phenyl-1, HPLC, UV detection, Methocarbamol, Guaifenesin B-Isomer, Guaifenesin, Guaiacol, Methocarbamol Isomer, Methocarbamol Dioxolone, Guaifenesin Impurity-D |
| Matrix: | Formulation |
| Author: | Manish Singh |
| Affiliation: | Thermo Fisher Scientific, APAC CoE Lab, Ahmedabad, India |
For Research Use Only. Not for use in diagnostic procedures.